Alfa Aesar™ 2-Methoxypyrazine, 98%

Manufacturer: Alfa Aesar™ A1029703

Quantity 1g

 View more versions of this product

Catalog No. AAA1029703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3149-28-8
    Molecular Formula C5H6N2O
    Formula Weight 110.12
    MDL Number MFCD00006126
    UN Number UN1993
    Beilstein 2241
    Chemical Name or Material 2-Methoxypyrazine

    Specifications

    Formula Weight 110.12
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C5H6N2O
    Boiling Point 147° to 149°C
    CAS 3149-28-8
    Beilstein 2241
    Health Hazard 1 Warning
    MDL Number MFCD00006126
    UN Number UN1993
    Quantity 1g
    Flash Point 45°C (113°F)
    TSCA TSCA
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 2-Methoxypyrazine
    Assay 98%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit