Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxypyridin-3-amine, 97%, Maybridge

Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
20265-38-7
C6H8N2O
124.143
MFCD00833386
YXFAOWYMDGUFIQ-UHFFFAOYSA-N
3-amino-2-methoxypyridine,2-methoxy-3-aminopyridine,2-methoxy-pyridin-3-ylamine,3-pyridinamine, 2-methoxy,2-methoxy-3-pyridylamine,2-methoxy-3-pyridinamine,2-methoxypyridine-3-amine,pubchem1269,2-methoxy pyridin-3-amine
2737488
2-methoxypyridin-3-amine
COC1=C(C=CC=N1)N
20265-38-7
C6H8N2O
124.143
MFCD00833386
YXFAOWYMDGUFIQ-UHFFFAOYSA-N
3-amino-2-methoxypyridine,2-methoxy-3-aminopyridine,2-methoxy-pyridin-3-ylamine,3-pyridinamine, 2-methoxy,2-methoxy-3-pyridylamine,2-methoxy-3-pyridinamine,2-methoxypyridine-3-amine,pubchem1269,2-methoxy pyridin-3-amine
2737488
2-methoxypyridin-3-amine
COC1=C(C=CC=N1)N
Packaging Amber Glass Bottle
Quantity 10g
CAS Min % 97.0
Assay Percent Range 97%
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 2-Methoxypyridin-3-amine

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning