Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxypyridine 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1628-89-3
C6H7NO
109.13
MFCD00006262
IWTFOFMTUOBLHG-UHFFFAOYSA-N
pyridine, 2-methoxy,2-methoxy pyridine,pyridine, methoxy,2-methoxy-pyridine,unii-geb38j8108,methyl 2-pyridyl ether,methoxypyridine,6-methoxypyridine,methyl pyridinyl ether,pubchem13201
74201
2-methoxypyridine
COC1=CC=CC=N1
1628-89-3
C6H7NO
109.13
MFCD00006262
IWTFOFMTUOBLHG-UHFFFAOYSA-N
pyridine, 2-methoxy,2-methoxy pyridine,pyridine, methoxy,2-methoxy-pyridine,unii-geb38j8108,methyl 2-pyridyl ether,methoxypyridine,6-methoxypyridine,methyl pyridinyl ether,pubchem13201
74201
2-methoxypyridine
COC1=CC=CC=N1
Boiling Point 142°C
Flash Point 32°C
Specific Gravity 1.04
Density 1.0400g/mL
Refractive Index 1.5010 to 1.503
Infrared Spectrum Authentic
Quantity 25g
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C5H4N)OCH3
Beilstein 21, 44
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,acetone and ether
Formula Weight 109.13
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Methoxypyridine, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-623-8

TSCA : TSCA