Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methoxythiazole, 98%

Manufacturer:  Alfa Aesar™ B2565514

Quantity 25g

 View more versions of this product

Catalog No. AAB2565514


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

14542-13-3
115.15
UN1993
C5H5NOS
MFCD01631143
2-Methoxythiazole

Specifications

14542-13-3
115.15
150° to 151°C
25g
2-Methoxythiazole
C5H5NOS
MFCD01631143
UN1993
49°C (120°F)
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit