Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1,3-cyclohexanedione, 98+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
200g
Packaging:
Glass bottle
1193-55-1
C7H10O2
126.15
MFCD00001587
VSGJHHIAMHUZKF-UHFFFAOYSA-N
2-methyl-1,3-cyclohexanedione,1,3-cyclohexanedione, 2-methyl,1,3-dimethylcycloadipic ketone,2-methyl-1,3-cyclohexandione,1-methyl-2,6-cyclohexanedione,ghl.pd_mitscher_leg0.98,2-methylcyclohexan-1,3-dion,2-methyl-cyclohexane-1,3-dione,pubchem15384,acmc-209a2g
70945
2-methylcyclohexane-1,3-dione
CC1C(=O)CCCC1=O
1193-55-1
C7H10O2
126.15
MFCD00001587
VSGJHHIAMHUZKF-UHFFFAOYSA-N
2-methyl-1,3-cyclohexanedione,1,3-cyclohexanedione, 2-methyl,1,3-dimethylcycloadipic ketone,2-methyl-1,3-cyclohexandione,1-methyl-2,6-cyclohexanedione,ghl.pd_mitscher_leg0.98,2-methylcyclohexan-1,3-dion,2-methyl-cyclohexane-1,3-dione,pubchem15384,acmc-209a2g
70945
2-methylcyclohexane-1,3-dione
CC1C(=O)CCCC1=O
Quantity 100g
Water 1% max. (K.F.)
Melting Point >204°C
Color Brown to White
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula CH3C6H7(=O)2
Beilstein 07, 558
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 126.15
Physical Form Powder, Crystals or Crystalline Powder
Percent Purity ≥98.0%
Chemical Name or Material 2-Methyl-1, 3-cyclohexanedione
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning