Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1,3-cyclopentanedione 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
765-69-5
C6H8O2
112.13
MFCD00001406
HXZILEQYFQYQCE-UHFFFAOYSA-N
2-methyl-1,3-cyclopentanedione,1,3-cyclopentanedione, 2-methyl,2-methyl-1,3-cyclopentadione,2-methyl-cyclopentane-1,3-dione,unii-c9lg5vp01c,c9lg5vp01c,methylcycle-d,pubchem2206,acmc-209p4n,benzil-related compound, 46
13005
2-methylcyclopentane-1,3-dione
CC1C(=O)CCC1=O
765-69-5
C6H8O2
112.13
MFCD00001406
HXZILEQYFQYQCE-UHFFFAOYSA-N
2-methyl-1,3-cyclopentanedione,1,3-cyclopentanedione, 2-methyl,2-methyl-1,3-cyclopentadione,2-methyl-cyclopentane-1,3-dione,unii-c9lg5vp01c,c9lg5vp01c,methylcycle-d,pubchem2206,acmc-209p4n,benzil-related compound, 46
13005
2-methylcyclopentane-1,3-dione
CC1C(=O)CCC1=O
Melting Point 212.0°C to 216.0°C
Color Beige to Tan
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 112.13
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Methyl-1, 3-cyclopentanedione, 98%
Show More Show Less

EINECSNumber : 212-153-2

TSCA : TSCA