Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1,3-thiazole-4-carboximidamide hydrochloride, 97%, Maybridge

Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
18876-82-9
C5H8ClN3S
177.65
MFCD00178765
YYLYRAUCGUIGIE-UHFFFAOYSA-N
2-methyl-1,3-thiazole-4-carboximidamide hydrochloride,2-methylthiazole-4-carboxamidine hydrochloride,2-methyl-thiazole-4-carboxamidine hydrochloride,2-methylthiazole-4-carboximidamide hydrochloride,2-methyl-1,3-thiazole-4-carboxamidine, chloride,4-thiazolecarboximidamide,2-methyl-,hydrochloride 1:1
2799402
2-methyl-1,3-thiazole-4-carboximidamide;hydrochloride
CC1=NC(=CS1)C(=N)N.Cl
18876-82-9
C5H8ClN3S
177.65
MFCD00178765
YYLYRAUCGUIGIE-UHFFFAOYSA-N
2-methyl-1,3-thiazole-4-carboximidamide hydrochloride,2-methylthiazole-4-carboxamidine hydrochloride,2-methyl-thiazole-4-carboxamidine hydrochloride,2-methylthiazole-4-carboximidamide hydrochloride,2-methyl-1,3-thiazole-4-carboxamidine, chloride,4-thiazolecarboximidamide,2-methyl-,hydrochloride 1:1
2799402
2-methyl-1,3-thiazole-4-carboximidamide;hydrochloride
CC1=NC(=CS1)C(=N)N.Cl
Quantity 5g
Packaging Amber Glass Bottle
CAS Min % 97.0
Assay Percent Range 97%
Formula Weight 177.66
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 3-thiazole-4-carboximidamide hydrochloride

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.