Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methyl-1,4-bis(trimethylsiloxy)benzene, 98%

Click to view available options
Quantity:
5g
25g
78018-53-8
C13H24O2Si2
268.503
MFCD02683083
IPYOHMJSEPFQSQ-UHFFFAOYSA-N
2-methyl-1,4-bis trimethylsiloxy benzene,trimethyl 3-methyl-4-trimethylsilyl oxy phenoxy silane,2,5-bis trimethylsiloxy toluene,methylhydroquinone, 2tms derivative,methylhydroquinone, bis trimethylsilyl ether,1,4-bis trimethylsilyl oxy-2-methylbenzene,benzene,2-methyl-1,4-bis trimethylsilyl oxy,2-methyl-1,4-phenylene bis oxy bis trimethylsilane
12691803
trimethyl-(2-methyl-4-trimethylsilyloxyphenoxy)silane
CC1=C(C=CC(=C1)O[Si](C)(C)C)O[Si](C)(C)C
78018-53-8
C13H24O2Si2
268.503
MFCD02683083
IPYOHMJSEPFQSQ-UHFFFAOYSA-N
2-methyl-1,4-bis trimethylsiloxy benzene,trimethyl 3-methyl-4-trimethylsilyl oxy phenoxy silane,2,5-bis trimethylsiloxy toluene,methylhydroquinone, 2tms derivative,methylhydroquinone, bis trimethylsilyl ether,1,4-bis trimethylsilyl oxy-2-methylbenzene,benzene,2-methyl-1,4-bis trimethylsilyl oxy,2-methyl-1,4-phenylene bis oxy bis trimethylsilane
12691803
trimethyl-(2-methyl-4-trimethylsilyloxyphenoxy)silane
CC1=C(C=CC(=C1)O[Si](C)(C)C)O[Si](C)(C)C
Formula Weight 268.51
Percent Purity 98%
Quantity 25g
Chemical Name or Material 2-Methyl-1,4-bis(trimethylsiloxy)benzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning