Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+/-)-2-Methyl-1-butanol, 98%

Click to view available options
Quantity:
100mL
500mL
2500mL
137-32-6
C5H12O
88.15
MFCD00004743
QPRQEDXDYOZYLA-UHFFFAOYSA-N
2-methyl-1-butanol,active amyl alcohol,1-butanol, 2-methyl,2-methylbutanol,sec-butylcarbinol,dl-2-methyl-1-butanol,2-methyl-n-butanol,2-methylbutyl alcohol,2-methyl butanol-1,primary active amyl alcohol
8723
CHEBI:48945
2-methylbutan-1-ol
CCC(C)CO
137-32-6
C5H12O
88.15
MFCD00004743
QPRQEDXDYOZYLA-UHFFFAOYSA-N
2-methyl-1-butanol,active amyl alcohol,1-butanol, 2-methyl,2-methylbutanol,sec-butylcarbinol,dl-2-methyl-1-butanol,2-methyl-n-butanol,2-methylbutyl alcohol,2-methyl butanol-1,primary active amyl alcohol
8723
CHEBI:48945
2-methylbutan-1-ol
CCC(C)CO
Applications
2-Methyl-1-butanol is a volatile metabolite produced by a number of different plant species. Used as a solvent in organic synthesis (introduction of active amyl group), in lubricants, plasticizers, additives for oils & paints. It is also employed as a perfuming agent.

Solubility
Soluble in water. [183 g/L (0 C)] and alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Quantity 500mL
Boiling Point 129°C to 131°C
Melting Point <-70°C
Flash Point 43°C (109°F)
UN Number UN1105
Beilstein 1718810
Merck Index 14,6030
Formula Weight 88.15
Percent Purity 98%
Chemical Name or Material (+/-)-2-Methyl-1-butanol

GHS H Statement
H226-H332-H335
Flammable liquid and vapour.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS

RTECSNumber : EL5250000

TSCA : TSCA