Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-buten-3-yne, 98%, Thermo Scientific Chemicals

Catalog Number AC336480250
Click to view available options
Quantity:
5 mL
10 mL
25 mL
50 mL
78-80-8
MFCD00008599
BOFLDKIFLIFLJA-UHFFFAOYSA-N
isopropenylacetylene, 2-methyl-1-buten-3-yne, valylene, 1-buten-3-yne, 2-methyl, 3-methyl-3-buten-1-yne, 2-methylbutenyne, isopropenyl acetylene, acmc-20anrd, 2-methyl-1-butenyne, 2-methyl-l-buten-3-yne
62323
2-methylbut-1-en-3-yne
CC(=C)C#C
78-80-8
MFCD00008599
BOFLDKIFLIFLJA-UHFFFAOYSA-N
isopropenylacetylene, 2-methyl-1-buten-3-yne, valylene, 1-buten-3-yne, 2-methyl, 3-methyl-3-buten-1-yne, 2-methylbutenyne, isopropenyl acetylene, acmc-20anrd, 2-methyl-1-butenyne, 2-methyl-l-buten-3-yne
62323
2-methylbut-1-en-3-yne
CC(=C)C#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Density 0.69g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C5H6
Refractive Index 1.4140 to 1.4180 (20°C, 589nm)
Quantity 25 mL
Molecular Weight (g/mol) 66.103
Formula Weight 66.1
Percent Purity ≥97.5% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-buten-3-yne
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Extremely flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information g

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit