Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-pentene 99%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
Packaging:
Glass bottle
763-29-1
C6H12
84.15
MFCD00009405
WWUVJRULCWHUSA-UHFFFAOYSA-N
2-methyl-1-pentene,2-methylpentene,1-pentene, 2-methyl,4-methyl-4-pentene,2-methyl-pentene-1,unii-okc25o38mc,okc25o38mc,acmc-1bixb,4-01-00-00841 beilstein handbook reference,c2h5ch2c ch3 =ch2
12986
2-methylpent-1-ene
CCCC(=C)C
763-29-1
C6H12
84.15
MFCD00009405
WWUVJRULCWHUSA-UHFFFAOYSA-N
2-methyl-1-pentene,2-methylpentene,1-pentene, 2-methyl,4-methyl-4-pentene,2-methyl-pentene-1,unii-okc25o38mc,okc25o38mc,acmc-1bixb,4-01-00-00841 beilstein handbook reference,c2h5ch2c ch3 =ch2
12986
2-methylpent-1-ene
CCCC(=C)C
Melting Point -136.0°C
Boiling Point 62.0°C
Density 0.6800g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3910 to 1.9300
Specific Gravity 0.68
Quantity 50mL
Color Colorless
Flash Point −26°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3CH2CH2C(CH3)=CH2
Beilstein 01, I, 90
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether
Formula Weight 84.15
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-Methyl-1-pentene, 99%
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Danger

EINECSNumber : 212-108-7

RTECSNumber : SB2230000

TSCA : TSCA