Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
38614-36-7,109-99-9
C4H7BrMg
159.31
FFUIUNZKXBJVSC-UHFFFAOYSA-M
2-methyl-1-propenylmagnesium bromide,2-methyl-1-propenyl magnesium bromide,2-methylpropenylmagnesium bromide,2-methyl-propenylmagnesium bromide,2-methylpropenyl magnesium bromide,bkviqodysrejqh-uhfffaoysa-m,2-methyl propenyl magnesium bromide,2-methylprop-1-enylmagnesium bromide,2-methylprop-1-enyl magnesium bromide
4456485
magnesium;2-methylprop-1-ene;bromide
CC(=[CH-])C.[Mg+2].[Br-]
38614-36-7,109-99-9
C4H7BrMg
159.31
FFUIUNZKXBJVSC-UHFFFAOYSA-M
2-methyl-1-propenylmagnesium bromide,2-methyl-1-propenyl magnesium bromide,2-methylpropenylmagnesium bromide,2-methyl-propenylmagnesium bromide,2-methylpropenyl magnesium bromide,bkviqodysrejqh-uhfffaoysa-m,2-methyl propenyl magnesium bromide,2-methylprop-1-enylmagnesium bromide,2-methylprop-1-enyl magnesium bromide
4456485
magnesium;2-methylprop-1-ene;bromide
CC(=[CH-])C.[Mg+2].[Br-]
Specific Gravity 0.952
Density 0.9520g/mL
Flash Point −15°C
Color Orange to Yellow
Quantity 100mL
Packaging AcroSeal™ Glass Bottle
CAS Min % 7.53,90.8
CAS Max % 9.2,92.47
Solubility Information Solubility in water: reacts
Formula Weight 159.31
Physical Form Solution
Chemical Name or Material 2-Methyl-1-propenylmagnesium bromide
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Suspected of causing cancer if inhaled.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flamma

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger