Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-pyrroline 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
50mL
Packaging:
Glass bottle
872-32-2
C5H9N
83.134
MFCD00128806
CTSZPNIMMLSKDV-UHFFFAOYSA-N
2-methyl-1-pyrroline,3,4-dihydro-5-methyl-2h-pyrrole,1pyrroline2methyl,2h-pyrrole, 3,4-dihydro-5-methyl,pubchem9412,2-methyl-1-pyrroline
70103
CHEBI:78856
5-methyl-3,4-dihydro-2H-pyrrole
CC1=NCCC1
872-32-2
C5H9N
83.134
MFCD00128806
CTSZPNIMMLSKDV-UHFFFAOYSA-N
2-methyl-1-pyrroline,3,4-dihydro-5-methyl-2h-pyrrole,1pyrroline2methyl,2h-pyrrole, 3,4-dihydro-5-methyl,pubchem9412,2-methyl-1-pyrroline
70103
CHEBI:78856
5-methyl-3,4-dihydro-2H-pyrrole
CC1=NCCC1
Color Beige to White
Refractive Index 1.4400 to 1.4420 (20°C, 589nm)
Quantity 50mL
Packaging Glass bottle
Density 0.8780g/mL
Infrared Spectrum Authentic
Water 2% max.
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 83.13
Physical Form Crystalline Powder and/or Chunks
Percent Purity ≥97.5% (GC)
Chemical Name or Material 2-Methyl-1-pyrroline, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 212-822-9

TSCA : TSCA