Alfa Aesar™ 2-Methyl-1-tetralone, 98%

Manufacturer: Alfa Aesar™ H3186509

Quantity 10g

 View more versions of this product

Catalog No. AAH3186509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1590-08-5
    Molecular Formula C11H12O
    Formula Weight 160.22
    MDL Number MFCD00001690
    Synonym 2-Methyl-1,2,3,4-tetrahydro-1-naphthalenone
    Chemical Name or Material 2-Methyl-1-tetralone

    Specifications

    Formula Weight 160.22
    MDL Number MFCD00001690
    Flash Point >110°C (230°F)
    Quantity 10g
    Molecular Formula C11H12O
    CAS 1590-08-5
    Synonym 2-Methyl-1,2,3,4-tetrahydro-1-naphthalenone
    Boiling Point 129°C (12mmHg)
    Chemical Name or Material 2-Methyl-1-tetralone
    Assay 98%

    Description

    Applications
    Methylation of 1-tetralone followed by the subsequent dehydrogenation of 2-methyl-1-tetralone in the presence of Pd/C catalyst also has been reported.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit