Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
1314216-34-6
C14H19BN2O2
258.13
CPQGLMCLTALRHF-UHFFFAOYSA-N
2-methyl-1h-benzimidazole-5-boronic acid pinacol ester,2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole,2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole,cpqglmcltalrhf-uhfffaoysa-n,2-methyl-1h-benzimidazole-5-boronic acidpinacol ester,2-methyl-1h-benzimidazole-5-boronic acid, pinacol ester,1h-benzimidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzo d imidazole,2-methyl-6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-benzo d imidazole,2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzimidazole
46738035
1314216-34-6
C14H19BN2O2
258.13
CPQGLMCLTALRHF-UHFFFAOYSA-N
2-methyl-1h-benzimidazole-5-boronic acid pinacol ester,2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole,2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole,cpqglmcltalrhf-uhfffaoysa-n,2-methyl-1h-benzimidazole-5-boronic acidpinacol ester,2-methyl-1h-benzimidazole-5-boronic acid, pinacol ester,1h-benzimidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzo d imidazole,2-methyl-6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-benzo d imidazole,2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzimidazole
46738035
Melting Point 211°C to 212°C
Quantity 5g
Infrared Spectrum Authentic
Color White to Yellow
NMR 96 % min. consistent with structure (1H-NMR)
Assay Percent Range 97%
Formula Weight 258.13
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning