Learn More
2-Methyl-2-butanol, 98%, Thermo Scientific Chemicals
Description
2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.
Solubility
Miscible with alcohol, ether, benzene, chloroform, glycerol, oils and acetone.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Specifications
Melting Point | -12°C |
Density | 0.805 g/mL |
Boiling Point | 101°C to 103°C |
Flash Point | 20°C (68°F) |
Assay Percent Range | 0.98 |
Refractive Index | 1.405 |
Quantity | 2500 mL |
UN Number | UN1105 |
Beilstein | 1361351 |
Merck Index | 14,7140 |
Show More |
Safety and Handling
Hazard Category | H225-H312+H332-H315-H318-H335-H336 |
Hazard Statement | GHS H Statement H225-H332-H315-H335 |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PENTANOLS |
EINECSNumber | 200-908-9 |
RTECSNumber | SC0175000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.