Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-imidazoline, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
534-26-9
C4H8N2
84.12
VWSLLSXLURJCDF-UHFFFAOYSA-N
2-methyl-2-imidazoline,2-methylimidazoline,1h-imidazole, 4,5-dihydro-2-methyl,methylglyoxalidine,2-imidazoline, 2-methyl,4,5-dihydro-2-methyl-1h-imidazole,unii-987f50e3pe,2-methyl imidazoline,2-methyl-4,5-dihydroimidazole,2-methyl-20imidazolin
10798
2-methyl-4,5-dihydro-1H-imidazole
CC1=NCCN1
534-26-9
C4H8N2
84.12
VWSLLSXLURJCDF-UHFFFAOYSA-N
2-methyl-2-imidazoline,2-methylimidazoline,1h-imidazole, 4,5-dihydro-2-methyl,methylglyoxalidine,2-imidazoline, 2-methyl,4,5-dihydro-2-methyl-1h-imidazole,unii-987f50e3pe,2-methyl imidazoline,2-methyl-4,5-dihydroimidazole,2-methyl-20imidazolin
10798
2-methyl-4,5-dihydro-1H-imidazole
CC1=NCCN1
Melting Point 104°C to 107°C
Boiling Point 110°C (2.0 Torr)
Quantity 5g
Color Colorless to Yellow or White to Yellow
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Merck Index 15,5696
Formula Weight 84.12
Physical Form Powder or Crystals or Needles
Percent Purity 95%
Chemical Name or Material 2-Methyl-2-imidazoline
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-596-6