Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methyl-2-nitro-1,3-propanediol, 97%

Click to view available options
Quantity:
5g
25g
100g
77-49-6
C4H9NO4
135.119
MFCD00024812
LOTYADDQWWVBDJ-UHFFFAOYSA-N
2-methyl-2-nitro-1,3-propanediol,nmpd,2-nitro-2-methyl-1,3-propanediol,nitroisobutylglycol,1,3-propanediol, 2-methyl-2-nitro,2-methyl-2-nitropropanediol,1,1-dimethylol-1-nitroethane,caswell no. 573n,epa pesticide chemical code 573900,acmc-1basy
6480
2-methyl-2-nitropropane-1,3-diol
CC(CO)(CO)[N+](=O)[O-]
77-49-6
C4H9NO4
135.119
MFCD00024812
LOTYADDQWWVBDJ-UHFFFAOYSA-N
2-methyl-2-nitro-1,3-propanediol,nmpd,2-nitro-2-methyl-1,3-propanediol,nitroisobutylglycol,1,3-propanediol, 2-methyl-2-nitro,2-methyl-2-nitropropanediol,1,1-dimethylol-1-nitroethane,caswell no. 573n,epa pesticide chemical code 573900,acmc-1basy
6480
2-methyl-2-nitropropane-1,3-diol
CC(CO)(CO)[N+](=O)[O-]
Quantity 25g
Melting Point 148°C to 151°C
Formula Weight 135.12
Percent Purity 97%
Chemical Name or Material 2-Methyl-2-nitro-1,3-propanediol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TY8925000

TSCA : TSCA