Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-nitropropane, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
594-70-7
C4H9NO2
103.12
MFCD00007393
AIMREYQYBFBEGQ-UHFFFAOYSA-N
propane, 2-methyl-2-nitro, trimethylnitromethane, 2-nitroisobutane, 2-nitro-2-methylpropane, tert-nitrobutane, 1,1-dimethyl-1-nitroethane, nitro-tert-butane, 2-methyl-2-nitro-propane, unii-9dkh5l679z, ccris 5044
11672
2-methyl-2-nitropropane
CC(C)(C)[N+](=O)[O-]
594-70-7
C4H9NO2
103.12
MFCD00007393
AIMREYQYBFBEGQ-UHFFFAOYSA-N
propane, 2-methyl-2-nitro, trimethylnitromethane, 2-nitroisobutane, 2-nitro-2-methylpropane, tert-nitrobutane, 1,1-dimethyl-1-nitroethane, nitro-tert-butane, 2-methyl-2-nitro-propane, unii-9dkh5l679z, ccris 5044
11672
2-methyl-2-nitropropane
CC(C)(C)[N+](=O)[O-]
Specific Gravity 0.95
Boiling Point 126°C to 127°C
Flash Point 19°C
Melting Point 24°C
Refractive Index 1.3990 to 1.403
Color White
Infrared Spectrum Authentic
Density 0.9500g/mL
Quantity 25g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5 % min. (GC)
Linear Formula (CH3)3CNO2
Beilstein 01, 129
Solubility Information Solubility in water: insoluble
Formula Weight 103.12
Physical Form Crystalline Powder, Crystals, or Flakes
Percent Purity 99%
Chemical Name or Material 2-Methyl-2-nitropropane, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-851-4

TSCA : TSCA