Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methyl-2-phenylacetamidoxime, 97%

Click to view available options
Quantity:
250mg
1g
925698-75-5
C9H12N2O
164.208
KFRRAHGTTHZCNG-UHFFFAOYSA-N
n-hydroxy-2-phenyl-propionamidine,z-n'-hydroxy-2-phenylpropanimidamide,2-phenyl-2-methylacetamidoxime
17889341
N'-hydroxy-2-phenylpropanimidamide
CC(C1=CC=CC=C1)C(=NO)N
925698-75-5
C9H12N2O
164.208
KFRRAHGTTHZCNG-UHFFFAOYSA-N
n-hydroxy-2-phenyl-propionamidine,z-n'-hydroxy-2-phenylpropanimidamide,2-phenyl-2-methylacetamidoxime
17889341
N'-hydroxy-2-phenylpropanimidamide
CC(C1=CC=CC=C1)C(=NO)N
Color Brown to White
Quantity 250mg
Formula Weight 164.2
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material 2-Methyl-2-phenylacetamidoxime