Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-2-Methyl-2-propanesulfinamide, 97%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
146374-27-8
C4H11NOS
121.198
MFCD05861480
CESUXLKAADQNTB-UHFFFAOYSA-N
2-methyl-2-propanesulfinamide,tert-butanesulfinamide,t-butylsulfinamide,tert-butylsulfinamide,1,1-dimethylethylsulfinamide,2-propanesulfinamide, 2-methyl,2-methyl-propane-2-sulfinic acid amide,tert-butylsulphinamide,unii-i85yc2za8o,i85yc2za8o
3382465
2-methylpropane-2-sulfinamide
CC(C)(C)S(=O)N
146374-27-8
C4H11NOS
121.198
MFCD05861480
CESUXLKAADQNTB-UHFFFAOYSA-N
2-methyl-2-propanesulfinamide,tert-butanesulfinamide,t-butylsulfinamide,tert-butylsulfinamide,1,1-dimethylethylsulfinamide,2-propanesulfinamide, 2-methyl,2-methyl-propane-2-sulfinic acid amide,tert-butylsulphinamide,unii-i85yc2za8o,i85yc2za8o
3382465
2-methylpropane-2-sulfinamide
CC(C)(C)S(=O)N
Applications
(±)-2-Methyl-2-propanesulfinamide is used in the synthesis of 4-sulfonyliminopiperidine and its reaction with benzyl Grignard to form an important pharmaceutical building block, 4-benzyl-4-aminopiperidne. It is also used to prepare spiro tosylaziridines via methylene insertion into the derived sulfonylimine.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Quantity 5g
Melting Point 97°C to 101°C
Formula Weight 121.2
Percent Purity 97%
Chemical Name or Material (±)-2-Methyl-2-propanesulfinamide