Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-propanethiol 99%, ACROS Organics™

Click to view available options
Quantity:
50mL
500mL
2.5L
Packaging:
Glass bottle
75-66-1
C4H10S
90.18
MFCD00004857
WMXCDAVJEZZYLT-UHFFFAOYSA-N
2-methyl-2-propanethiol,tert-butyl mercaptan,tert-butylthiol,tert-butanethiol,2-propanethiol, 2-methyl,tert-butylmercaptan,t-butyl mercaptan,t-butylmercaptan,2-isobutanethiol,tertiary-butyl mercaptan
6387
2-methylpropane-2-thiol
CC(C)(C)S
75-66-1
C4H10S
90.18
MFCD00004857
WMXCDAVJEZZYLT-UHFFFAOYSA-N
2-methyl-2-propanethiol,tert-butyl mercaptan,tert-butylthiol,tert-butanethiol,2-propanethiol, 2-methyl,tert-butylmercaptan,t-butyl mercaptan,t-butylmercaptan,2-isobutanethiol,tertiary-butyl mercaptan
6387
2-methylpropane-2-thiol
CC(C)(C)S
Refractive Index 1.4220 to 1.424
Melting Point -1.1°C
Boiling Point 62°C to 65°C
Flash Point -24°C
Viscosity 0.638 mPa.s (20°C)
Specific Gravity 0.8
Quantity 2.5L
Packaging Glass bottle
Density 0.8000g/mL
Infrared Spectrum Authentic
Color Colorless
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)3CSH
Beilstein 01, 383
Merck Index 15, 1582
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in liquid h2s,very soluble in alcohol and ether
Formula Weight 90.18
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2-Methyl-2-propanethiol, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 200-890-2

RTECSNumber : TZ7660000

TSCA : TSCA