Alfa Aesar™ 2-(Methyl-2-pyridylamino)ethanol, 98%

Manufacturer: Alfa Aesar™ B2555518

Quantity 50g

 View more versions of this product

Catalog No. AAB2555518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 122321-04-4
    Molecular Formula C8H12N2O
    Formula Weight 152.19
    MDL Number MFCD02667633
    Chemical Name or Material 2-(Methyl-2-pyridylamino)ethanol

    Specifications

    Formula Weight 152.19
    Health Hazard 1 Danger
    Quantity 50g
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C8H12N2O
    MDL Number MFCD02667633
    CAS 122321-04-4
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 2-(Methyl-2-pyridylamino)ethanol
    Assay 98%

    Description

    Applications
    2-Methyl-2-pyridylaminoethanol is used as a reagent to synthesize Rosiglitazone (Maleate: R693500), a thiazolinedione drug used to reduce insulin resistance in patients with Type 2 diabetes.

    Solubility
    Soluble in water.

    Notes
    Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit