Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Methyl-2-pyridylamino)ethanol, 98%

Manufacturer:  Alfa Aesar™ B2555530

Quantity 250g

 View more versions of this product

Catalog No. AAB2555530


  Check Availability
Add to cart

Specifications

Description

Applications
2-Methyl-2-pyridylaminoethanol is used as a reagent to synthesize Rosiglitazone (Maleate: R693500), a thiazolinedione drug used to reduce insulin resistance in patients with Type 2 diabetes.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

122321-04-4
152.19
2-(Methyl-2-pyridylamino)ethanol
C8H12N2O
MFCD02667633

Specifications

122321-04-4
152.19
250g
98%
C8H12N2O
MFCD02667633
2-(Methyl-2-pyridylamino)ethanol

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit