Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-thiopseudourea hemisulfate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
867-44-7
C2H6N21/2H2SO4
139.18
MFCD00013055
BZZXQZOBAUXLHZ-UHFFFAOYSA-N
s-methylisothiourea hemisulfate,2-methyl-2-thiopseudourea hemisulfate,carbamimidothioic acid, methyl ester, sulfate 2:1,s-methylisothiourea sulfate 2:1,s-methylthiouronium sulfate 2:1,bis 2-methylisothiouronium sulphate,methyl carbamimidothioate sulfate 2:1,methylcarbamimidothioate sulfate 2:1,s-methylisothiourea, sulfate,2-methyl-2-thiopseudourea sulfate 2:1
13347
methyl carbamimidothioate;sulfuric acid
CSC(=N)N.CSC(=N)N.OS(=O)(=O)O
867-44-7
C2H6N21/2H2SO4
139.18
MFCD00013055
BZZXQZOBAUXLHZ-UHFFFAOYSA-N
s-methylisothiourea hemisulfate,2-methyl-2-thiopseudourea hemisulfate,carbamimidothioic acid, methyl ester, sulfate 2:1,s-methylisothiourea sulfate 2:1,s-methylthiouronium sulfate 2:1,bis 2-methylisothiouronium sulphate,methyl carbamimidothioate sulfate 2:1,methylcarbamimidothioate sulfate 2:1,s-methylisothiourea, sulfate,2-methyl-2-thiopseudourea sulfate 2:1
13347
methyl carbamimidothioate;sulfuric acid
CSC(=N)N.CSC(=N)N.OS(=O)(=O)O
Melting Point 240.0°C to 241.0°C
Packaging Plastic bottle
Infrared Spectrum Authentic
Color White
Quantity 500g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula [CH3SC(=NH)NH2]2·1/2H2SO4
Solubility Information Solubility in water: 260g/L (20°C). Other solubilities: 4g/L methanol (20°C)
Formula Weight 139.18
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Methyl-2-thiopseudourea hemisulfate, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-759-7

RTECSNumber : UM9200000

TSCA : TSCA