Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-vinyloxirane, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
1838-94-4
C5H8O
84.12
FVCDMHWSPLRYAB-UHFFFAOYSA-N
2-methyl-2-vinyloxirane,isoprene monoxide,ccris 5685,3,4-epoxy-3-methyl-1-butene,2-methyl-2-vinyl-oxirane,oxirane, 2-ethenyl-2-methyl-, 2r,acmc-20m5m5,oxirane, 2-ethenyl-2-methyl,4,4,4-3fmet-1-2-furyl-1,3-butandione
92166
2-ethenyl-2-methyloxirane
CC1(CO1)C=C
1838-94-4
C5H8O
84.12
FVCDMHWSPLRYAB-UHFFFAOYSA-N
2-methyl-2-vinyloxirane,isoprene monoxide,ccris 5685,3,4-epoxy-3-methyl-1-butene,2-methyl-2-vinyl-oxirane,oxirane, 2-ethenyl-2-methyl-, 2r,acmc-20m5m5,oxirane, 2-ethenyl-2-methyl,4,4,4-3fmet-1-2-furyl-1,3-butandione
92166
2-ethenyl-2-methyloxirane
CC1(CO1)C=C
Infrared Spectrum Authentic
Quantity 5g
Density 0.8000g/mL
Color Colorless
Flash Point −8°C
Specific Gravity 0.8
Boiling Point 80.0°C to 81.0°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 84.12
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 2-Methyl-2-vinyloxirane
Show More Show Less

GHS H Statement
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with pl

GHS Signal Word: Danger