Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2H-indazole-6-boronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1001907-57-8
C8H9BN2O2
175.98
MFCD09870057
JADBVQWCMHPPQA-UHFFFAOYSA-N
2-methyl-2h-indazole-6-boronic acid,2-methyl-2h-indazol-6-yl boronic acid,2-methylindazole-6-boronic acid,6-borono-2-methyl-2h-indazole,6-boronic acid-2-methyl-2h-indazole,boronicacid, b-2-methyl-2h-indazol-6-yl,b-2-methyl-2h-indazol-6-yl-boronic acid,boronic acid, b-2-methyl-2h-indazol-6-yl,acmc-2097na,boron-mol bm794
44630579
(2-methylindazol-6-yl)boronic acid
B(C1=CC2=NN(C=C2C=C1)C)(O)O
1001907-57-8
C8H9BN2O2
175.98
MFCD09870057
JADBVQWCMHPPQA-UHFFFAOYSA-N
2-methyl-2h-indazole-6-boronic acid,2-methyl-2h-indazol-6-yl boronic acid,2-methylindazole-6-boronic acid,6-borono-2-methyl-2h-indazole,6-boronic acid-2-methyl-2h-indazole,boronicacid, b-2-methyl-2h-indazol-6-yl,b-2-methyl-2h-indazol-6-yl-boronic acid,boronic acid, b-2-methyl-2h-indazol-6-yl,acmc-2097na,boron-mol bm794
44630579
(2-methylindazol-6-yl)boronic acid
B(C1=CC2=NN(C=C2C=C1)C)(O)O
Color White
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 306.0°C to 311.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 175.98
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Methyl-2H-indazole-6-boronic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning