Alfa Aesar™ 2-Methyl-3-(2-furyl)propenal, 97%

Manufacturer: Alfa Aesar™ A1012530

Quantity 250g

 View more versions of this product

Catalog No. AAA1012530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 874-66-8
    Molecular Formula C8H8O2
    Formula Weight 136.15
    MDL Number MFCD00063240
    Beilstein 2438
    Synonym 2-Methyl-3-(2-furyl)acrolein
    Chemical Name or Material 2-Methyl-3- (2-furyl)propenal

    Specifications

    Formula Weight 136.15
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 874-66-8
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number LT8530600
    Molecular Formula C8H8O2
    MDL Number MFCD00063240
    Beilstein 2438
    Health Hazard 1 Warning
    Quantity 250g
    Flash Point >100°C (212°F)
    Recommended Storage 2°C to 8°C
    TSCA TSCA
    Synonym 2-Methyl-3-(2-furyl)acrolein
    Boiling Point 280° to 281°C
    Chemical Name or Material 2-Methyl-3- (2-furyl)propenal
    Assay 97%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit