Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-nitroaniline, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
603-83-8
C7H8N2O2
152.15
MFCD00007731
HFCFJYRLBAANKN-UHFFFAOYSA-N
2-amino-6-nitrotoluene,3-nitro-o-toluidine,benzenamine, 2-methyl-3-nitro,o-toluidine, 3-nitro,2-methyl-3-nitro-aniline,1-amino-2-methyl-3-nitrobenzene,2-nitro-6-aminotoluene,2-methyl-3-nitrobenzenamine,2-methyl-3-nitro-phenylamine,2-methyl-3-nitrophenyl amine
11783
2-methyl-3-nitroaniline
CC1=C(C=CC=C1[N+](=O)[O-])N
603-83-8
C7H8N2O2
152.15
MFCD00007731
HFCFJYRLBAANKN-UHFFFAOYSA-N
2-amino-6-nitrotoluene,3-nitro-o-toluidine,benzenamine, 2-methyl-3-nitro,o-toluidine, 3-nitro,2-methyl-3-nitro-aniline,1-amino-2-methyl-3-nitrobenzene,2-nitro-6-aminotoluene,2-methyl-3-nitrobenzenamine,2-methyl-3-nitro-phenylamine,2-methyl-3-nitrophenyl amine
11783
2-methyl-3-nitroaniline
CC1=C(C=CC=C1[N+](=O)[O-])N
Boiling Point 305°C
Infrared Spectrum Authentic
Quantity 100g
Color Green-Yellow to Yellow
Packaging Glass bottle
Melting Point 87°C to 91°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3C6H3(NO2)NH2
Beilstein 12, 848
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in ethanol and ether
Formula Weight 152.15
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Methyl-3-nitroaniline
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger