Alfa Aesar™ 2-Methyl-3-nitroaniline, 98%

Manufacturer: Alfa Aesar™ A1034714

Quantity 25g

 View more versions of this product

Catalog No. AAA1034714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 603-83-8
    Molecular Formula C7H8N2O2
    Formula Weight 152.15
    MDL Number MFCD00007731
    UN Number UN2660
    Beilstein 388393
    Synonym 2-Amino-6-nitrotoluene; 3-Nitro-o-toluidine
    Chemical Name or Material 2-Methyl-3-nitroaniline

    Specifications

    Formula Weight 152.15
    Health Hazard 3 GHS P Statement
    P280h-P273-P309-P310-P302+P352
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Molecular Formula C7H8N2O2
    CAS 603-83-8
    Beilstein 388393
    Synonym 2-Amino-6-nitrotoluene; 3-Nitro-o-toluidine
    Health Hazard 1 Danger
    Boiling Point 305°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROTOLUIDINES (MONO)
    UN Number UN2660
    Odor Characteristic
    RTECS Number XU8200000
    MDL Number MFCD00007731
    Melting Point 88° to 91°C
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H373
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Chemical Name or Material 2-Methyl-3-nitroaniline
    Assay 98%

    GHS H Statement
    H301-H311-H331-H373
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P280h-P273-P309-P310-P302+P352
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit