Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-nitroanisole, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
4837-88-1
C8H9NO3
167.16
HQCZLEAGIOIIMC-UHFFFAOYSA-N
2-methyl-3-nitroanisole,2-methoxy-6-nitrotoluene,benzene, 1-methoxy-2-methyl-3-nitro,1-methoxy-2-methyl-3-nitro-benzene,anisole, 2-methyl-3-nitro,6-methoxy-2-nitrotoluene,1-meo-2-me-3-no2-benzene,3-nitro-2-methyl anisole,3-methoxy-2-methyl-1-nitrobenzene,2-methyl-3-nitroanisole 2-methoxy-6-nitrotoluene
78554
1-methoxy-2-methyl-3-nitrobenzene
CC1=C(C=CC=C1OC)[N+](=O)[O-]
4837-88-1
C8H9NO3
167.16
HQCZLEAGIOIIMC-UHFFFAOYSA-N
2-methyl-3-nitroanisole,2-methoxy-6-nitrotoluene,benzene, 1-methoxy-2-methyl-3-nitro,1-methoxy-2-methyl-3-nitro-benzene,anisole, 2-methyl-3-nitro,6-methoxy-2-nitrotoluene,1-meo-2-me-3-no2-benzene,3-nitro-2-methyl anisole,3-methoxy-2-methyl-1-nitrobenzene,2-methyl-3-nitroanisole 2-methoxy-6-nitrotoluene
78554
1-methoxy-2-methyl-3-nitrobenzene
CC1=C(C=CC=C1OC)[N+](=O)[O-]
Packaging Glass Bottle
Melting Point 54.0°C to 55.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Color Brown to Yellow
Quantity 1g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3C6H3(NO2)OCH3
Beilstein 06,I,178
Formula Weight 167.16
Physical Form Powder or Crystalline Powder
Percent Purity 99%
Chemical Name or Material 2-Methyl-3-nitroanisole
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-424-5