Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methyl-3-nitrophenylacetic acid, 96%

Click to view available options
Quantity:
5g
25g
100g
23876-15-5
C9H9NO4
195.174
MFCD07782132
LWIOFILTAJJDLA-UHFFFAOYSA-N
2-methyl-3-nitrophenylacetic acid,2-methyl-3-nitrophenyl acetic acid,2-2-methyl-3-nitrophenyl acetic acid,2-methyl-3-nitro-benzeneacetic acid,3-nitro-o-tolyl acetic acid,benzeneacetic acid, 2-methyl-3-nitro,2-methyl-3-nitrophenylaceticacid,2-methyl-3-nitrobenzeneacetic acid,2-methyl-3-nitro phenyl acetic acid,pubchem18230
13530897
CC1=C(C=CC=C1[N+](=O)[O-])CC(=O)O
23876-15-5
C9H9NO4
195.174
MFCD07782132
LWIOFILTAJJDLA-UHFFFAOYSA-N
2-methyl-3-nitrophenylacetic acid,2-methyl-3-nitrophenyl acetic acid,2-2-methyl-3-nitrophenyl acetic acid,2-methyl-3-nitro-benzeneacetic acid,3-nitro-o-tolyl acetic acid,benzeneacetic acid, 2-methyl-3-nitro,2-methyl-3-nitrophenylaceticacid,2-methyl-3-nitrobenzeneacetic acid,2-methyl-3-nitro phenyl acetic acid,pubchem18230
13530897
CC1=C(C=CC=C1[N+](=O)[O-])CC(=O)O
Quantity 100g
Melting Point 132°C to 136°C
Beilstein 2581973
Formula Weight 195.17
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 2-Methyl-3-nitrophenylacetic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C