Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-4(5)-nitroimidazole 99.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
696-23-1
C4H5N3O2
127.103
MFCD00005191
FFYTTYVSDVWNMY-UHFFFAOYSA-N
2-methyl-5-nitroimidazole,2-methyl-4-nitroimidazole,2-methyl-4-nitro-1h-imidazole,menidazole,1h-imidazole, 2-methyl-4-nitro,1h-imidazole, 2-methyl-5-nitro,2-methyl-4 5-nitroimidazole,imidazole, 2-methyl-4-nitro,unii-24ag2ww15w,imidazole, 2-methyl-5-nitro
12760
2-methyl-5-nitro-1H-imidazole
CC1=NC=C(N1)[N+](=O)[O-]
696-23-1
C4H5N3O2
127.103
MFCD00005191
FFYTTYVSDVWNMY-UHFFFAOYSA-N
2-methyl-5-nitroimidazole,2-methyl-4-nitroimidazole,2-methyl-4-nitro-1h-imidazole,menidazole,1h-imidazole, 2-methyl-4-nitro,1h-imidazole, 2-methyl-5-nitro,2-methyl-4 5-nitroimidazole,imidazole, 2-methyl-4-nitro,unii-24ag2ww15w,imidazole, 2-methyl-5-nitro
12760
2-methyl-5-nitro-1H-imidazole
CC1=NC=C(N1)[N+](=O)[O-]
Melting Point 253°C
Color Yellow
Quantity 25g
Formula Weight 127.10
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (T)
Chemical Name or Material 2-Methyl-4(5)-nitroimidazole

RTECSNumber : NI7550000

TSCA : Yes