Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-4-isothiazolin-3-one, 50% aqueous solution

Catalog No. p-7077826
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 2682-20-4 | C4H5NOS | 115.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2682-20-4
Molecular Formula C4H5NOS
Molecular Weight (g/mol) 115.15
InChI Key BEGLCMHJXHIJLR-UHFFFAOYSA-N
Synonym 2-methyl-4-isothiazolin-3-one, methylisothiazolinone, 2-methyl-4-isothiazoline-3-one, 2-methylisothiazol-3 2h-one, 2-methyl-3 2h-isothiazolone, n-methylisothiazolone, 3 2h-isothiazolone, 2-methyl, caswell no. 572a, 2-methyl-3-isothiazolone, unii-229d0e1qfa
PubChem CID 39800
ChEBI CHEBI:53620
IUPAC Name 2-methyl-1,2-thiazol-3-one
SMILES CN1C(=O)C=CS1

Specifications

Density 1.1600g/mL
pH 5.0 to 7.0 (1% aq. soln.)
Quantity 5 g
Specific Gravity 1.16
Formula Weight 115.15
Chemical Name or Material 2-Methyl-4-isothiazolin-3-one
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic if inhaled.
May cause respiratory irritation.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 220-239-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.