Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-4-isothiazolin-3-one, 50% aqueous solution, Acros Organics

Click to view available options
Quantity:
5g
25g
2682-20-4
C4H5NOS
115.15
BEGLCMHJXHIJLR-UHFFFAOYSA-N
2-methyl-4-isothiazolin-3-one,methylisothiazolinone,2-methyl-4-isothiazoline-3-one,2-methylisothiazol-3 2h-one,2-methyl-3 2h-isothiazolone,n-methylisothiazolone,3 2h-isothiazolone, 2-methyl,caswell no. 572a,2-methyl-3-isothiazolone,unii-229d0e1qfa
39800
CHEBI:53620
2-methyl-1,2-thiazol-3-one
CN1C(=O)C=CS1
2682-20-4
C4H5NOS
115.15
BEGLCMHJXHIJLR-UHFFFAOYSA-N
2-methyl-4-isothiazolin-3-one,methylisothiazolinone,2-methyl-4-isothiazoline-3-one,2-methylisothiazol-3 2h-one,2-methyl-3 2h-isothiazolone,n-methylisothiazolone,3 2h-isothiazolone, 2-methyl,caswell no. 572a,2-methyl-3-isothiazolone,unii-229d0e1qfa
39800
CHEBI:53620
2-methyl-1,2-thiazol-3-one
CN1C(=O)C=CS1
Quantity 25g
pH 5.0 to 7.0 (1% aq. soln.)
Density 1.1600g/mL
Specific Gravity 1.16
Formula Weight 115.15
Chemical Name or Material 2-Methyl-4-isothiazolin-3-one

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic if inhaled.
May cause respiratory irritation.
Very toxic to aquatic life.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

EINECSNumber : 220-239-6