Alfa Aesar™ 2'-Methyl-4'-nitroacetanilide, 97%

Manufacturer: Alfa Aesar™ B2205414

Quantity 25g

 View more versions of this product

Catalog No. AAB2205414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2719-15-5
    Molecular Formula C9H10N2O3
    Formula Weight 194.19
    MDL Number MFCD00033896
    Chemical Name or Material 2'-Methyl-4'-nitroacetanilide

    Specifications

    Formula Weight 194.19
    Quantity 25g
    TSCA TSCA
    Molecular Formula C9H10N2O3
    MDL Number MFCD00033896
    CAS 2719-15-5
    Health Hazard 1 Warning
    Melting Point 198° to 200°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2'-Methyl-4'-nitroacetanilide
    Assay 97%

    Description

    Applications
    2 ft.-Methyl-4 ft.-nitroacetanilide acts as the reactant in the synthesis of TNT isomers.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, bases.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit