Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methyl-4-nitroaniline, 98%

Click to view available options
Quantity:
500g
2500g
99-52-5
C7H8N2O2
152.15
MFCD00007734
XTTIQGSLJBWVIV-UHFFFAOYSA-N
2-amino-5-nitrotoluene, 4-nitro-o-toluidine, 5-nitro-2-aminotoluene, p-nitro-o-toluidine, benzenamine, 2-methyl-4-nitro, fast red rl base, 1-amino-2-methyl-4-nitrobenzene, diabase red rl, ansibases red rl, devol red rl
7441
2-methyl-4-nitroaniline
CC1=CC(=CC=C1N)[N+]([O-])=O
99-52-5
C7H8N2O2
152.15
MFCD00007734
XTTIQGSLJBWVIV-UHFFFAOYSA-N
2-amino-5-nitrotoluene, 4-nitro-o-toluidine, 5-nitro-2-aminotoluene, p-nitro-o-toluidine, benzenamine, 2-methyl-4-nitro, fast red rl base, 1-amino-2-methyl-4-nitrobenzene, diabase red rl, ansibases red rl, devol red rl
7441
2-methyl-4-nitroaniline
CC1=CC(=CC=C1N)[N+]([O-])=O
Odor Characteristic
Melting Point 130°C to 133°C
Quantity 500g
UN Number UN2660
Beilstein 775772
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 2-Methyl-4-nitroaniline

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c

H301+H311+H331-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROTOLUIDINES (MONO)

EINECSNumber : 202-762-1

RTECSNumber : XU8210000

TSCA : Yes

Recommended Storage : Ambient temperatures