Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methyl-4-(trifluoromethoxy)aniline, 97+%

Click to view available options
Quantity:
1g
5g
86256-59-9
C8H8F3NO
191.153
MFCD01631541
IIDBMILLZRYZCH-UHFFFAOYSA-N
2-methyl-4-trifluoromethoxy aniline,2-methyl-4-trifluoromethoxy benzenamine,4-trifluoromethoxy-2-methylaniline,4-trifluoromethoxy-o-toluidine,benzenamine, 2-methyl-4-trifluoromethoxy,2-methyl-4-trifluoromethoxy phenylamine,pubchem8512,ksc495s8h,2-amino-5-trifluoromethoxy toluene
2775546
2-methyl-4-(trifluoromethoxy)aniline
CC1=C(C=CC(=C1)OC(F)(F)F)N
86256-59-9
C8H8F3NO
191.153
MFCD01631541
IIDBMILLZRYZCH-UHFFFAOYSA-N
2-methyl-4-trifluoromethoxy aniline,2-methyl-4-trifluoromethoxy benzenamine,4-trifluoromethoxy-2-methylaniline,4-trifluoromethoxy-o-toluidine,benzenamine, 2-methyl-4-trifluoromethoxy,2-methyl-4-trifluoromethoxy phenylamine,pubchem8512,ksc495s8h,2-amino-5-trifluoromethoxy toluene
2775546
2-methyl-4-(trifluoromethoxy)aniline
CC1=C(C=CC(=C1)OC(F)(F)F)N
Quantity 5g
UN Number UN2810
Formula Weight 191.15
Percent Purity ≥97%
Chemical Name or Material 2-Methyl-4-(trifluoromethoxy)aniline

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.