Alfa Aesar™ 2-Methyl-4-trimethylsilyl-1-buten-3-yne, 97%

Manufacturer: Alfa Aesar™ 4418918

Quantity 50g

 View more versions of this product

Catalog No. AA4418918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18387-60-5
    Molecular Formula (CH3)3SiC≡CC(CH3)=CH2
    Formula Weight 138.28
    MDL Number MFCD00190206
    UN Number UN1993
    Chemical Name or Material 2-Methyl-4-trimethylsilyl-1-buten-3-yne

    Specifications

    Formula Weight 138.28
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    CAS 18387-60-5
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    MDL Number MFCD00190206
    Boiling Point 32°C (15mmHg)
    Quantity 50g
    Density 0.778g/mL
    Molecular Formula (CH3)3SiC≡CC(CH3)=CH2
    UN Number UN1993
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material 2-Methyl-4-trimethylsilyl-1-buten-3-yne
    Assay 97%

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit