Alfa Aesar™ 2-Methyl-5-nitro-2H-indazole, 98+%

Manufacturer: Alfa Aesar™ B2236006

Quantity 5g

 View more versions of this product

Catalog No. AAB2236006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5228-48-8
    Molecular Formula C8H7N3O2
    Formula Weight 177.16
    MDL Number MFCD00051809
    Beilstein 167044
    Chemical Name or Material 2-Methyl-5-nitro-2H-indazole

    Specifications

    Formula Weight 177.16
    Molecular Formula C8H7N3O2
    MDL Number MFCD00051809
    CAS 5228-48-8
    Beilstein 167044
    Melting Point 126° to 132°C
    Quantity 5g
    Chemical Name or Material 2-Methyl-5-nitro-2H-indazole
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit