Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methyl-5-nitro-2H-indazole, 98+%

Click to view available options
Quantity:
0.25g
1g
5g
5228-48-8
C8H7N3O2
177.163
MFCD00051809
IWPBWVZEOVCZRX-UHFFFAOYSA-N
2-methyl-5-nitro-2h-indazole,2h-indazole, 2-methyl-5-nitro,2-methyl-5-nitro-1h-indazole,2-methyl-5-nitro-indazole,acmc-1b10q,2h-indazole,2-methyl-5-nitro,2h-benzopyrazole, 2-methyl-5-nitro
97550
2-methyl-5-nitroindazole
CN1C=C2C=C(C=CC2=N1)[N+](=O)[O-]
5228-48-8
C8H7N3O2
177.163
MFCD00051809
IWPBWVZEOVCZRX-UHFFFAOYSA-N
2-methyl-5-nitro-2h-indazole,2h-indazole, 2-methyl-5-nitro,2-methyl-5-nitro-1h-indazole,2-methyl-5-nitro-indazole,acmc-1b10q,2h-indazole,2-methyl-5-nitro,2h-benzopyrazole, 2-methyl-5-nitro
97550
2-methyl-5-nitroindazole
CN1C=C2C=C(C=CC2=N1)[N+](=O)[O-]
Quantity 1g
Melting Point 126°C to 132°C
Beilstein 167044
Formula Weight 177.16
Percent Purity ≥98%
Chemical Name or Material 2-Methyl-5-nitro-2H-indazole