Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methyl-5-nitrobenzotrifluoride, 98%

Click to view available options
Quantity:
1g
5g
25g
89976-12-5
C8H6F3NO2
205.136
MFCD01631684
SVQCVQCIZWSPPX-UHFFFAOYSA-N
2-methyl-5-nitrobenzotrifluoride,1-methyl-4-nitro-2-trifluoromethyl benzene,2-trifluoromethyl-4-nitrotoluene,4-nitro-2-trifluoromethyl toluene,1-methyl-4-nitro-2-trifluoromethyl-benzene,1-methyl-4-nitro-2-trifluoromethyl-benzene,benzene, 1-methyl-4-nitro-2-trifluoromethyl,3-trifluoromethyl-4-methylnitrobenzene,2-trifluoromethyl-1-methyl-4-nitrobenzene,pubchem4444
2775447
1-methyl-4-nitro-2-(trifluoromethyl)benzene
CC1=C(C=C(C=C1)[N+](=O)[O-])C(F)(F)F
89976-12-5
C8H6F3NO2
205.136
MFCD01631684
SVQCVQCIZWSPPX-UHFFFAOYSA-N
2-methyl-5-nitrobenzotrifluoride,1-methyl-4-nitro-2-trifluoromethyl benzene,2-trifluoromethyl-4-nitrotoluene,4-nitro-2-trifluoromethyl toluene,1-methyl-4-nitro-2-trifluoromethyl-benzene,1-methyl-4-nitro-2-trifluoromethyl-benzene,benzene, 1-methyl-4-nitro-2-trifluoromethyl,3-trifluoromethyl-4-methylnitrobenzene,2-trifluoromethyl-1-methyl-4-nitrobenzene,pubchem4444
2775447
1-methyl-4-nitro-2-(trifluoromethyl)benzene
CC1=C(C=C(C=C1)[N+](=O)[O-])C(F)(F)F
Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
Color White
Sensitivity Moisture Sensitive
Quantity 5g
UN Number UN2811
Beilstein 2506013
Formula Weight 205.14
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Methyl-5-nitrobenzotrifluoride

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.