Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-5-nitrophenylboronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
957062-84-9
C13H18BNO4
263.1
LYFUUPOOXAXEEP-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-2-methyl-5-nitrophenyl-1,3,2-dioxaborolane, 2-methyl-5-nitrophenylboronic acid pinacol ester, 2-methyl-5-nitrophenylboronic acid, pinacol ester, 2-methyl-5-nitrophenyl boric acid pinacol ester, 3-hydroxy-2,3-dimethylbutan-2-yl hydrogen 2-methyl-5-nitrophenylboronate, pubchem18757, 2-methyl-5-nitrophenylboronicacidpinacolester, 2-methyl-5-nitrobenzeneboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-2-methyl-5-nitro-phenyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-methyl-5-nitrophenyl
17750269
4,4,5,5-tetramethyl-2-(2-methyl-5-nitrophenyl)-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)[N+](=O)[O-])C
957062-84-9
C13H18BNO4
263.1
LYFUUPOOXAXEEP-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-2-methyl-5-nitrophenyl-1,3,2-dioxaborolane, 2-methyl-5-nitrophenylboronic acid pinacol ester, 2-methyl-5-nitrophenylboronic acid, pinacol ester, 2-methyl-5-nitrophenyl boric acid pinacol ester, 3-hydroxy-2,3-dimethylbutan-2-yl hydrogen 2-methyl-5-nitrophenylboronate, pubchem18757, 2-methyl-5-nitrophenylboronicacidpinacolester, 2-methyl-5-nitrobenzeneboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-2-methyl-5-nitro-phenyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-methyl-5-nitrophenyl
17750269
4,4,5,5-tetramethyl-2-(2-methyl-5-nitrophenyl)-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)[N+](=O)[O-])C
Packaging Glass Bottle
Melting Point 90.0°C to 92.0°C
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 263.1
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-nitrophenylboronic acid pinacol ester

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning