Alfa Aesar™ 2-Methyl-5-(pentafluorothio)aniline, 97%

Manufacturer: Alfa Aesar™ H33782MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH33782MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C7H8F5NS
    Formula Weight 233.2
    MDL Number MFCD16652447
    UN Number UN2811
    Chemical Name or Material 2-Methyl-5-(pentafluorothio)aniline

    Specifications

    Formula Weight 233.2
    Molecular Formula C7H8F5NS
    MDL Number MFCD16652447
    UN Number UN2811
    Health Hazard 1 Danger
    Quantity 250mg
    Health Hazard 2 GHS H Statement
    H301-H311-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 2-Methyl-5-(pentafluorothio)aniline
    Assay 97%

    GHS H Statement
    H301-H311-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit