Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-5H-dibenz[b,f]azepine-5-carboxamide p.a., Acros Organics

Click to view available options
Quantity:
5mg
70401-32-0
C16H14N2O
250.301
MFCD00074838
WLUGVRXHDCWYLA-UHFFFAOYSA-N
2-methyl-5h-dibenz b,f azepine-5-carboxamide,2-methyl-5h-dibenz b,f azepine-5-carboxamide,5h-dibenz b,f azepine-5-carboxamide,2-methyl,2-methyldibenzo b,f azepine-5-carboxamide,2-methyl-5 h-dibenz b,f azepine-5-carboxamide,2-methyl-5h-dibenzo b,f azepine-5-carboxamide,3-methylbenzo b 1 benzazepine-11-carboxamide,6-methyl-2-azatricyclo 9.4.0.0 3 ,? pentadeca-1 11 ,3 8 ,4,6,9,12,14-heptaene-2-carboxamide,6-methyl-2-azatricyclo 9.4.0.0 3 ,? pentadeca-1 11 ,3,5,7,9,12,14-heptaene-2-carboxamide,6-methyl-2-azatricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3,5,7,9,11,13-heptaene-2-carboxamide
719579
3-methylbenzo[b][1]benzazepine-11-carboxamide
CC1=CC2=C(C=C1)N(C3=CC=CC=C3C=C2)C(=O)N
70401-32-0
C16H14N2O
250.301
MFCD00074838
WLUGVRXHDCWYLA-UHFFFAOYSA-N
2-methyl-5h-dibenz b,f azepine-5-carboxamide,2-methyl-5h-dibenz b,f azepine-5-carboxamide,5h-dibenz b,f azepine-5-carboxamide,2-methyl,2-methyldibenzo b,f azepine-5-carboxamide,2-methyl-5 h-dibenz b,f azepine-5-carboxamide,2-methyl-5h-dibenzo b,f azepine-5-carboxamide,3-methylbenzo b 1 benzazepine-11-carboxamide,6-methyl-2-azatricyclo 9.4.0.0 3 ,? pentadeca-1 11 ,3 8 ,4,6,9,12,14-heptaene-2-carboxamide,6-methyl-2-azatricyclo 9.4.0.0 3 ,? pentadeca-1 11 ,3,5,7,9,12,14-heptaene-2-carboxamide,6-methyl-2-azatricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3,5,7,9,11,13-heptaene-2-carboxamide
719579
3-methylbenzo[b][1]benzazepine-11-carboxamide
CC1=CC2=C(C=C1)N(C3=CC=CC=C3C=C2)C(=O)N
Color White
Water Contains water in varying amounts, 10% max. (amount specified in each lot)
Infrared Spectrum Authentic
Quantity 5mg
CAS Min % 99.0
CAS Max % 100.0
Fieser 01,1045
Formula Weight 250.3
Physical Form Fine Powder
Percent Purity ≥99%
Chemical Name or Material 2-Methyl-5H-dibenz[b, f]azepine-5-carboxamide, p.a.
Show More Show Less