Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methyl-6-nitrobenzoxazole, 98%

Click to view available options
Quantity:
1g
5g
25g
5683-43-2
C8H6N2O3
178.147
MFCD00022848
DPVZLXWUFYMLBK-UHFFFAOYSA-N
2-methyl-6-nitrobenzoxazole,benzoxazole, 2-methyl-6-nitro,acmc-1azgy,2-methyl-6-nitro-benzoxazole,benzoxazole,2-methyl-6-nitro,2-methyl-6-nitrobenzo d oxazole
79766
2-methyl-6-nitro-1,3-benzoxazole
CC1=NC2=C(O1)C=C(C=C2)[N+](=O)[O-]
5683-43-2
C8H6N2O3
178.147
MFCD00022848
DPVZLXWUFYMLBK-UHFFFAOYSA-N
2-methyl-6-nitrobenzoxazole,benzoxazole, 2-methyl-6-nitro,acmc-1azgy,2-methyl-6-nitro-benzoxazole,benzoxazole,2-methyl-6-nitro,2-methyl-6-nitrobenzo d oxazole
79766
2-methyl-6-nitro-1,3-benzoxazole
CC1=NC2=C(O1)C=C(C=C2)[N+](=O)[O-]
Applications
2-Methyl-6-nitrobenzoxazole is used as a flavoring agents.

Solubility
Soluble in oils. Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Quantity 1g
Melting Point 154°C to 156°C
Formula Weight 146.17
Percent Purity 98%
Chemical Name or Material 2-Methyl-6-nitrobenzoxazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning