Alfa Aesar™ 2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride, 97%

Manufacturer: Alfa Aesar™ H34468MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH34468MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C7H5ClF3NO2S
    Formula Weight 259.64
    MDL Number MFCD14583150
    UN Number UN3261
    Chemical Name or Material 2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride

    Specifications

    Formula Weight 259.64
    UN Number UN3261
    Health Hazard 1 Danger
    Melting Point 57° to 59°C
    Quantity 250mg
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage −30° to −10°C
    Molecular Formula C7H5ClF3NO2S
    MDL Number MFCD14583150
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride
    Assay 97%

    Description

    Applications
    Use as chiral derivatizing agents in the determination of the enantiomeric excess of alcohols and amines. They are widely used as important intermediates in the synthesis of dyes, drugs and pigments.

    Notes
    Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture. Water, heat.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    −30° to −10°C

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit