Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
1257535-05-9
C7H5ClF3NO2S
259.64
BNCNLCKWRCXBQH-UHFFFAOYSA-N
2-methyl-6-trifluoromethyl pyridine-3-sulfonyl chloride,3-chlorosulphonyl-2-methyl-6-trifluoromethyl pyridine
51342031
2-methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride
CC1=C(C=CC(=N1)C(F)(F)F)S(=O)(=O)Cl
1257535-05-9
C7H5ClF3NO2S
259.64
BNCNLCKWRCXBQH-UHFFFAOYSA-N
2-methyl-6-trifluoromethyl pyridine-3-sulfonyl chloride,3-chlorosulphonyl-2-methyl-6-trifluoromethyl pyridine
51342031
2-methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride
CC1=C(C=CC(=N1)C(F)(F)F)S(=O)(=O)Cl
Packaging Glass Bottle
Quantity 1g
NMR Consistent with structure (1H−NMR)
Infrared Spectrum Authentic
Assay Percent Range 97%
Formula Weight 259.64
Percent Purity 97%
Chemical Name or Material 2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin wi

GHS Signal Word: Danger