Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methyl-D-phenylalanine, 95%

Click to view available options
Quantity:
250mg
1g
5g
80126-54-1
C10H13NO2
179.219
MFCD01860870
NHBKDLSKDKUGSB-SECBINFHSA-N
d-2-methylphenylalanine,r-2-amino-3-o-tolyl propanoic acid,2-methyl-d-phenylalanine,2r-2-amino-3-2-methylphenyl propanoic acid,d-2-methylphe,d-2-me-phe-oh,d-phenylalanine, 2-methyl,o-methyl-d-phenylalanine,h-d-phe 2-me-oh,2-methy-d-phenylalanine
6951464
(2R)-2-amino-3-(2-methylphenyl)propanoic acid
CC1=CC=CC=C1CC(C(=O)O)N
80126-54-1
C10H13NO2
179.219
MFCD01860870
NHBKDLSKDKUGSB-SECBINFHSA-N
d-2-methylphenylalanine,r-2-amino-3-o-tolyl propanoic acid,2-methyl-d-phenylalanine,2r-2-amino-3-2-methylphenyl propanoic acid,d-2-methylphe,d-2-me-phe-oh,d-phenylalanine, 2-methyl,o-methyl-d-phenylalanine,h-d-phe 2-me-oh,2-methy-d-phenylalanine
6951464
(2R)-2-amino-3-(2-methylphenyl)propanoic acid
CC1=CC=CC=C1CC(C(=O)O)N
Beilstein 6175785
Formula Weight 179.22
Physical Form Powder
Percent Purity 95%
Color White
Quantity 250mg
Chemical Name or Material 2-Methyl-D-phenylalanine

storageNote1 : 2° to 8°C