Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methyl-L-phenylalanine, 97%

$138.01 - $439.01

Chemical Identifiers

CAS 80126-53-0
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD00044971
Synonym H-Phe(2-Me)-OH
Chemical Name or Material 2-Methyl-L-phenylalanine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH27663ME SDS Alfa Aesar™
H27663ME
500mg Each for $439.01
Check Availability
Add to cart
AAH27663MC SDS Alfa Aesar™
H27663MC
100mg Each for $138.01
Check Availability
Add to cart
Specifications Description & Specifications

Chemical Identifiers

80126-53-0
179.22
H-Phe(2-Me)-OH
C10H13NO2
MFCD00044971
2-Methyl-L-phenylalanine

Specifications

80126-53-0
179.22
H-Phe(2-Me)-OH
97%
C10H13NO2
MFCD00044971
2-Methyl-L-phenylalanine
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit